siRNA Details
| Gene Symbol: SLC2A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001290.1 | NP_001001290.1 | 13 | 1551 | ||
| NM_020041.2 | NP_064425.2 | 12 | 1540 | |||
| XM_006713968.3 | XP_006714031.1 | 12 | 1652 | |||
| XM_011513859.2 | XP_011512161.1 | 12 | 1647 | |||
| XM_011513860.2 | XP_011512162.1 | 12 | 1647 | |||
| XM_017008457.1 | XP_016863946.1 | 12 | 1665 | |||
| XM_017008460.1 | XP_016863949.1 | 11 | 3197 | |||
| GenBank mRNA | AF210317.1 | - | 12 | 1533 | ||
| BC018897.2 | - | 13 | 1551 | 3949549 | ||
| BC110414.1 | - | 12 | 1570 | 5762360 | ||

SLC2A9
Genomic Map