siRNA Details
| Gene Symbol: PNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002687.3 | NP_002678.2 | 2 | 262 | ||
| GenBank mRNA | AK223612.1 | - | 2 | 197 | ||
| AK292579.1 | - | 2 | 246 | |||
| BC062602.1 | - | 2 | 173 | 5491466 | ||
| CD687183.1 | - | 2 | 270 | |||
| U77718.1 | - | 2 | 195 | |||
| Y09703.1 | - | 2 | 173 | |||

PNN
Genomic Map