siRNA Details
| Gene Symbol: GLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000169.2 | NP_000160.1 | 3 | 508 | ||
| GenBank mRNA | AK222627.1 | - | 3 | 420 | ||
| AK291095.1 | - | 3 | 427 | |||
| AK297148.1 | - | 3 | 420 | |||
| BC002689.2 | - | 3 | 404 | 3609235 | ||
| BT006864.1 | - | 3 | 398 | |||
| D00039.1 | - | 3 | 322 | |||
| X05790.1 | - | 3 | 421 | |||
| X16889.1 | - | 3 | 400 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_013463, AK040081, AK054547, AK145011, AK145148, and others.

GLA
Genomic Map