siRNA Details
Gene Symbol: SGSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000199.3 | NP_000190.1 | 4 | 563 | ||
XM_005257582.3 | XP_005257639.1 | 4 | 496 | |||
XM_005257583.4 | XP_005257640.1 | 4 | 496 | |||
XM_011525126.1 | XP_011523428.1 | 4 | 496 | |||
XM_011525127.2 | XP_011523429.1 | 4 | 496 | |||
XM_017024951.1 | XP_016880440.1 | 4 | 496 | |||
XM_017024952.1 | XP_016880441.1 | 4 | 496 | |||
GenBank mRNA | AB209900.1 | - | 4 | 485 | ||
AK095969.1 | - | 3 | 417 | |||
AK222890.1 | - | 4 | 496 | |||
AK291257.1 | - | 4 | 496 | |||
AK304706.1 | - | 4 | 536 | |||
AK309927.1 | - | 4 | 514 | |||
BC047318.1 | - | 4 | 488 | 5226903 | ||
U30894.1 | - | 4 | 488 |
