siRNA Details
| Gene Symbol: DIO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000793.5 | NP_000784.2 | 4 | 1711 | ||
| NM_001007023.4 | NP_001007024.1 | 3 | 1588 | |||
| NM_001242502.2 | NP_001229431.1 | 4 | 1722 | |||
| NM_001242503.2 | NP_001229432.1 | 3 | 1614 | |||
| NM_001324462.1 | NP_001311391.1 | 3 | 1727 | |||
| NM_013989.4 | NP_054644.1 | 2 | 1480 | |||
| GenBank mRNA | AF093774.1 | - | 2 | 1480 | ||
| BC063118.1 | - | 2 | 1363 | |||
| BC136514.1 | - | 2 | 1350 | 9020504 | ||
| U53506.1 | - | 2 | 1459 | |||

DIO2
Genomic Map