siRNA Details
| Gene Symbol: DIO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001362.3 | NP_001353.4 | 1 | 1189 | ||
| GenBank mRNA | AK292310.1 | - | 1 | 1045 | ||
| BC017717.1 | - | 1 | 1037 | 4395664 | ||
| BX161470.1 | - | 1 | 1189 | |||
| S79854.1 | - | 1 | 1184 | |||

DIO3
Genomic Map