siRNA Details
| Gene Symbol: ARSD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001669.3 | NP_001660.2 | 5 | 931 | ||
| NM_009589.3 | NP_033667.2 | 5 | 931 | |||
| XM_005274514.2 | XP_005274571.1 | 5 | 901 | |||
| XM_005274515.2 | XP_005274572.1 | 5 | 901 | |||
| GenBank mRNA | AF160499.1 | - | 5 | 831 | ||
| AK314479.1 | - | 5 | 895 | |||
| BC020229.1 | - | 859 | ||||
| X83572.1 | - | 5 | 907 | |||

ARSD
Genomic Map