siRNA Details
| Gene Symbol: CHI3L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025197.1 | NP_001020368.1 | 4 | 410 | ||
| NM_001025199.1 | NP_001020370.1 | 4 | 470 | |||
| NM_004000.2 | NP_003991.2 | 5 | 440 | |||
| GenBank mRNA | AK223316.1 | - | 4 | 350 | ||
| AK298466.1 | - | 4 | 361 | |||
| BC011460.1 | - | 5 | 378 | 4214121 | ||
| BG698401.1 | - | 5 | 440 | |||
| BM787868.1 | - | 4 | 460 | |||
| BT006767.1 | - | 5 | 369 | |||
| U49835.1 | - | 5 | 389 | |||
| U58514.1 | - | 5 | 405 | |||
| U58515.1 | - | 4 | 471 | |||

CHI3L2
Genomic Map