siRNA Details
| Gene Symbol: NIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185093.1 | NP_001172022.1 | 6 | 1386 | ||
| NM_001185094.1 | NP_001172023.1 | 7 | 1155 | |||
| NM_005600.2 | NP_005591.1 | 7 | 1154 | |||
| XM_005245214.2 | XP_005245271.1 | 7 | 1505 | |||
| XM_005245215.4 | XP_005245272.1 | 7 | 1166 | |||
| XM_005245216.3 | XP_005245273.1 | 7 | 1177 | |||
| GenBank mRNA | AF069987.1 | - | 7 | 1125 | ||

NIT1
Genomic Map