siRNA Details
| Gene Symbol: DPYSL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006426.2 | NP_006417.2 | 14 | 1819 | ||
| XM_005252658.3 | XP_005252715.1 | 14 | 1808 | |||
| XM_011539125.2 | XP_011537427.1 | 15 | 1822 | |||
| XM_017015482.1 | XP_016870971.1 | 14 | 3083 | |||
| XM_017015483.1 | XP_016870972.1 | 15 | 3236 | |||
| GenBank mRNA | AB006713.1 | - | 14 | 1813 | ||
| BC029280.1 | - | 14 | 1703 | |||
| BC136329.1 | - | 14 | 1688 | 9020316 | ||
| BC136330.1 | - | 14 | 1688 | 9020317 | ||

DPYSL4
Genomic Map