siRNA Details
| Gene Symbol: PADI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013358.2 | NP_037490.2 | 2 | 347 | ||
| XM_011541307.1 | XP_011539609.1 | 2 | 350 | |||
| XM_017001101.1 | XP_016856590.1 | 2 | 250 | |||
| XM_017001102.1 | XP_016856591.1 | 2 | 250 | |||
| XM_017001103.1 | XP_016856592.1 | 2 | 350 | |||
| GenBank mRNA | AB033768.2 | - | 2 | 338 | ||
| AK026652.1 | - | 2 | 349 | |||
| AK225541.1 | - | 2 | 349 | |||
| AK314644.1 | - | 2 | 348 | |||
| BC130574.1 | - | 2 | 326 | 40146605 | ||
| BC136402.1 | - | 2 | 326 | 9020389 | ||

PADI1
Genomic Map