siRNA Details
| Gene Symbol: VNN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291702.1 | NP_001278631.1 | 1 | 177 | ||
| NM_001291703.1 | NP_001278632.1 | 1 | 177 | |||
| GenBank mRNA | AB435052.1 | - | 1 | 104 | ||
| AB435053.1 | - | 1 | 104 | |||
| AB435054.1 | - | 1 | 104 | |||
| AB435055.1 | - | 1 | 104 | |||
| AB435056.1 | - | 1 | 104 | |||
| AB435057.1 | - | 1 | 104 | |||
| AB435058.1 | - | 1 | 104 | |||
| AB435059.1 | - | 1 | 104 | |||
| AB435060.1 | - | 1 | 104 | |||
| AB435061.1 | - | 1 | 104 | |||
| AJ238982.1 | - | 1 | 149 | |||
| AK075110.1 | - | 1 | 176 | |||
| AK314708.1 | - | 1 | 177 | |||
| BC104416.2 | - | 1 | 164 | |||
| BC104417.1 | - | 1 | 164 | |||
| BC113022.1 | - | 1 | 164 | |||
| BC113023.1 | - | 1 | 164 | |||
| BC148317.2 | - | 1 | 164 | 40015223 | ||
| DA670661.1 | - | 1 | 178 | |||

VNN3
Genomic Map