siRNA Details
| Gene Symbol: ART4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021071.2 | NP_066549.2 | 2 | 944 | ||
| GenBank mRNA | AF290204.1 | - | 2 | 677 | ||
| AK291662.1 | - | 2 | 960 | |||
| AY829434.1 | - | 2 | 913 | |||
| AY829437.1 | - | 2 | 527 | |||
| BC074727.2 | - | 2 | 614 | 30915180 | ||
| BX096478.1 | - | 1 | 332 | |||

ART4
Genomic Map