siRNA Details
| Gene Symbol: CDK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258.2 | NP_001249.1 | 4 | 309 | ||
| GenBank mRNA | DB455174.1 | - | 4 | 310 | ||
| X66357.1 | - | 3 | 321 | |||
| Gene Symbol: TEN1-CDK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037709.1 | 6 | 2069 | |||
| GenBank mRNA | AK097104.1 | - | 6 | 2026 | ||
| BX647274.1 | - | 6 | 2055 | |||

CDK3
Genomic Map