siRNA Details
| Gene Symbol: PSMB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002793.3 | NP_002784.1 | 1, 2 | 207 | ||
| GenBank mRNA | AB209078.1 | - | 1, 2 | 137 | ||
| AB451312.1 | - | 1, 2 | 120 | |||
| AB451442.1 | - | 1, 2 | 120 | |||
| AK058050.1 | - | 1, 2 | 177 | |||
| AK223193.1 | - | 1, 2 | 158 | |||
| BC000508.2 | - | 1, 2 | 158 | 2822268 | ||
| BC020807.1 | - | 1, 2 | 158 | 4281426 | ||
| BP369175.1 | - | 1, 2 | 219 | |||
| BT019720.1 | - | 1, 2 | 120 | |||
| D00761.1 | - | 1, 2 | 132 | |||

PSMB1
Genomic Map