siRNA Details
| Gene Symbol: SCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005063.4 | NP_005054.3 | 6 | 2324 | ||
| GenBank mRNA | AB032261.1 | - | 6 | 2204 | ||
| AB208982.1 | - | 6 | 2081 | |||
| AF097514.1 | - | 6 | 2070 | |||
| AF109362.1 | - | 1 | 48 | |||
| BU940035.1 | - | 1 | 333 | |||

SCD
Genomic Map