siRNA Details
| Gene Symbol: PGLS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012088.2 | NP_036220.1 | 2, 3 | 445 | ||
| XM_011527897.2 | XP_011526199.1 | 2, 3 | 1045 | |||
| GenBank mRNA | AF091091.1 | - | 1, 2 | 545 | ||
| AJ243972.1 | - | 2, 3 | 412 | |||
| BC014006.2 | - | 2, 3 | 419 | 4053022 | ||
| BG764228.1 | - | 2, 3 | 446 | |||
| GQ472229.1 | - | 2, 3 | 419 | |||

PGLS
Genomic Map