siRNA Details
| Gene Symbol: SLC9A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF073299.1 | - | 9 | 1870 | ||
| BC136377.1 | - | 9 | 1940 | 9020364 | ||
| BC136378.1 | - | 9 | 1940 | 9020365 | ||
| S83549.1 | - | 3 | 253 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC9A2| Gene Symbol: SLC9A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF073299.1 | - | 9 | 1870 | ||
| BC136377.1 | - | 9 | 1940 | 9020364 | ||
| BC136378.1 | - | 9 | 1940 | 9020365 | ||
| S83549.1 | - | 3 | 253 | |||