siRNA Details
| Gene Symbol: SLC35C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281457.1 | NP_001268386.1 | 10 | 2072 | ||
| NM_001281458.1 | NP_001268387.1 | 11 | 2011 | |||
| NM_001281459.1 | NP_001268388.1 | 10 | 2009 | |||
| NM_001281460.1 | NP_001268389.1 | 11 | 2475 | |||
| NM_015945.11 | NP_057029.8 | 10 | 2143 | |||
| NM_173073.3 | NP_775096.1 | 10 | 2080 | |||
| NM_173179.3 | NP_775271.1 | 10 | 2338 | |||

SLC35C2
Genomic Map