siRNA Details
| Gene Symbol: SLC4A1AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018158.2 | NP_060628.2 | 3 | 1379 | ||
| GenBank mRNA | AK001486.1 | - | 3 | 1379 | ||
| AK300958.1 | - | 5 | 889 | |||
| AY028435.1 | - | 3 | 1355 | |||
| AY117688.1 | - | 3 | 768 | |||
| BC098302.1 | - | 3 | 1374 | 40026925 | ||
| BC098358.1 | - | 3 | 1374 | 40026922 | ||
| BC099711.1 | - | 3 | 1374 | 40026923 | ||
| BC099739.1 | - | 3 | 1372 | 40026926 | ||

SLC4A1AP
Genomic Map