siRNA Details
| Gene Symbol: PLIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145311.1 | NP_001138783.1 | 7 | 1039 | ||
| NM_002666.4 | NP_002657.3 | 7 | 1110 | |||
| XM_005254934.4 | XP_005254991.1 | 7 | 968 | |||
| GenBank mRNA | AB005293.1 | - | 7 | 1053 | ||
| BC031084.1 | - | 7 | 1053 | 5284402 | ||

PLIN1
Genomic Map