siRNA Details
| Gene Symbol: MORC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085354.2 | NP_001078823.1 | 15 | 2552 | ||
| NM_024657.4 | NP_078933.3 | 15 | 2552 | |||
| XM_005262190.3 | XP_005262247.1 | 14 | 2442 | |||
| XM_006724691.2 | XP_006724754.1 | 13 | 2186 | |||
| XM_011531027.2 | XP_011529329.1 | 12 | 1840 | |||
| XM_017029844.1 | XP_016885333.1 | 14 | 2442 | |||
| GenBank mRNA | AK021627.1 | - | 4 | 866 | ||
| AK056235.1 | - | 11 | 1764 | |||
| AK300305.1 | - | 14 | 2127 | |||
| EF125209.1 | - | 15 | 2277 | |||
| EF125210.1 | - | 14 | 2336 | |||

MORC4
Genomic Map