siRNA Details
| Gene Symbol: SLC26A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000112.3 | NP_000103.2 | 3 | 1303 | ||
| XM_017009191.1 | XP_016864680.1 | 3 | 1311 | |||
| GenBank mRNA | AK290358.1 | - | 3 | 1126 | ||
| AK312596.1 | - | 3 | 1254 | |||
| BC059390.1 | - | 3 | 1157 | 30341764 | ||
| U14528.1 | - | 2 | 1062 | |||

SLC26A2
Genomic Map