siRNA Details
| Gene Symbol: SLC20A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257180.1 | NP_001244109.1 | 2 | 930 | ||
| NM_001257181.1 | NP_001244110.1 | 2 | 561 | |||
| NM_006749.4 | NP_006740.1 | 2 | 893 | |||
| XM_005273613.3 | XP_005273670.1 | 2 | 668 | |||
| XM_005273615.3 | XP_005273672.1 | 2 | 668 | |||
| XM_006716390.3 | XP_006716453.1 | 2 | 668 | |||
| XM_006716391.3 | XP_006716454.1 | 2 | 4013 | |||
| XM_017013748.1 | XP_016869237.1 | 2 | 784 | |||
| XM_017013749.1 | XP_016869238.1 | 2 | 4013 | |||
| XM_017013750.1 | XP_016869239.1 | 2 | 668 | |||
| XM_017013751.1 | XP_016869240.1 | 2 | 542 | |||
| XM_017013752.1 | XP_016869241.1 | 2 | 784 | |||
| GenBank mRNA | AK291202.1 | - | 2 | 930 | ||
| AK314774.1 | - | 2 | 562 | |||
| BC028600.2 | - | 2 | 605 | 4820601 | ||
| L20852.1 | - | 1 | 479 | |||

SLC20A2
Genomic Map