siRNA Details
| Gene Symbol: SLC30A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013309.5 | NP_037441.2 | 5 | 1104 | ||
| XM_011521997.2 | XP_011520299.1 | 5 | 1608 | |||
| XM_017022560.1 | XP_016878049.1 | 4 | 910 | |||
| GenBank mRNA | AF025409.1 | - | 4 | 792 | ||
| AK290874.1 | - | 4 | 910 | |||
| AY029482.1 | - | 1 | 69 | |||
| BC026089.2 | - | 5 | 1104 | 4829839 | ||

SLC30A4
Genomic Map