siRNA Details
| Gene Symbol: SLC18A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003055.2 | NP_003046.2 | 1 | 1380 | ||
| GenBank mRNA | AK313094.1 | - | 1 | 1269 | ||
| BC007765.2 | - | 1 | 1306 | 4136084 | ||
| U09210.1 | - | 1 | 1382 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC18A3| Gene Symbol: SLC18A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003055.2 | NP_003046.2 | 1 | 1380 | ||
| GenBank mRNA | AK313094.1 | - | 1 | 1269 | ||
| BC007765.2 | - | 1 | 1306 | 4136084 | ||
| U09210.1 | - | 1 | 1382 | |||