siRNA Details
| Gene Symbol: SLC30A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318949.1 | NP_001305878.1 | 5, 6 | 704 | ||
| NM_001318950.1 | NP_001305879.1 | 5, 6 | 849 | |||
| NM_001318951.1 | NP_001305880.1 | 4, 5 | 631 | |||
| NM_003459.4 | NP_003450.2 | 4, 5 | 767 | |||
| GenBank mRNA | AK313600.1 | - | 4, 5 | 722 | ||
| BC028358.2 | - | 4, 5 | 755 | 4837595 | ||
| U76010.1 | - | 4, 5 | 664 | |||

SLC30A3
Genomic Map