siRNA Details
| Gene Symbol: ATP2B4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001396.2 | NP_001001396.1 | 18 | 3779 | ||
| NM_001684.4 | NP_001675.3 | 18 | 3779 | |||
| GenBank mRNA | AK026443.1 | - | 6 | 805 | ||
| AK292458.1 | - | 17 | 3118 | |||
| BC140774.1 | - | 18 | 3451 | 9021642 | ||
| BC150293.1 | - | 17 | 3136 | 8860318 | ||
| BX537444.1 | - | 18 | 4006 | |||
| CR749393.1 | - | 18 | 3709 | |||
| CR936708.1 | - | 18 | 3734 | |||
| M25874.1 | - | 17 | 3279 | |||
| M83363.1 | - | 17 | 3279 | |||

ATP2B4
Genomic Map