siRNA Details
| Gene Symbol: PSG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130014.1 | NP_001123486.1 | 4 | 1028 | ||
| NM_002781.3 | NP_002772.3 | 4 | 1028 | |||
| XM_017027003.1 | XP_016882492.1 | 4 | 1026 | |||
| GenBank mRNA | AA151017.1 | - | 2 | 296 | ||
| BC012607.1 | - | 4 | 995 | 4043912 | ||
| BG621332.1 | - | 2 | 449 | |||
| L36607.1 | - | 3 | 664 | |||
| M25384.1 | - | 5 | 996 | |||
| M73713.1 | - | 4 | 982 | |||

PSG5
Genomic Map