siRNA Details
| Gene Symbol: IP6K3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142883.1 | NP_001136355.1 | 5 | 1039 | ||
| NM_054111.4 | NP_473452.2 | 4 | 910 | |||
| XM_005248842.3 | XP_005248899.1 | 5 | 819 | |||
| XM_005248843.3 | XP_005248900.1 | 3 | 930 | |||
| XM_011514295.2 | XP_011512597.1 | 4 | 922 | |||
| GenBank mRNA | AF393812.1 | - | 3 | 605 | ||
| AK056586.2 | - | 4 | 829 | |||
| AK223572.1 | - | 4 | 833 | |||
| AK289935.1 | - | 5 | 959 | |||
| BC089389.1 | - | 4 | 910 | 30530548 | ||

IP6K3
Genomic Map