siRNA Details
| Gene Symbol: UBE2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204880.1 | NP_001191809.1 | 6 | 1764 | ||
| NM_003338.4 | NP_003329.1 | 7 | 1796 | |||
| XM_017016609.1 | XP_016872098.1 | 7 | 1695 | |||
| GenBank mRNA | BM468116.1 | - | 1 | 500 | ||

UBE2D1
Genomic Map