siRNA Details
| Gene Symbol: UBE2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003339.2 | NP_003330.1 | 7 | 1311 | ||
| NM_181838.1 | NP_862821.1 | 8 | 1475 | |||
| XM_017009820.1 | XP_016865309.1 | 10 | 1682 | |||
| GenBank mRNA | AF317220.1 | - | 5 | 619 | ||
| AK001311.1 | - | 7 | 1084 | |||
| AK001428.1 | - | 7 | 1138 | |||
| AK310273.1 | - | 7 | 1040 | |||
| AY651263.1 | - | 7 | 1067 | |||
| BC019052.2 | - | 7 | 1095 | |||
| BC033349.1 | - | 7 | 1311 | 4827210 | ||

UBE2D2
Genomic Map