siRNA Details
| Gene Symbol: UBE2H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001202498.1 | NP_001189427.1 | 7 | 612 | ||
| NM_003344.3 | NP_003335.1 | 7 | 1008 | |||
| NM_182697.2 | NP_874356.1 | 5 | 915 | |||
| GenBank mRNA | AK091088.1 | - | 1 | 956 | ||
| AL832212.1 | - | 1 | 1017 | |||
| BC006277.2 | - | 7 | 920 | 3957107 | ||
| BI088146.1 | - | 7 | 614 | |||
| Z29328.1 | - | 7 | 656 | |||
| Z29330.1 | - | 7 | 656 | |||
| Z29331.1 | - | 2 | 273 | |||

UBE2H
Genomic Map