siRNA Details
| Gene Symbol: SUCLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003850.2 | NP_003841.1 | 4 | 556 | ||
| XM_017020806.1 | XP_016876295.1 | 4 | 776 | |||
| XM_017020807.1 | XP_016876296.1 | 6 | 791 | |||
| XM_017020808.1 | XP_016876297.1 | 4 | 543 | |||
| XM_017020809.1 | XP_016876298.1 | 5 | 640 | |||
| GenBank mRNA | AB035863.1 | - | 4 | 556 | ||
| AF058953.1 | - | 3 | 388 | |||
| AF087890.1 | - | 4 | 564 | |||
| AK001458.1 | - | 4 | 454 | |||
| AK001847.1 | - | 4 | 543 | |||
| AK225452.1 | - | 4 | 543 | |||
| AK299437.1 | - | 4 | 495 | |||
| AK302311.1 | - | 5 | 615 | |||
| AK315513.1 | - | 4 | 518 | |||
| AK316271.1 | - | 5 | 871 | |||
| BC027587.2 | - | 4 | 508 | 4829291 | ||
| BX640602.1 | - | 4 | 527 | |||
| KU178442.1 | - | 4 | 499 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011506, AK088801, AF058955, AK150594, BC057605, and others.

SUCLA2
Genomic Map