siRNA Details
| Gene Symbol: PSMB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270481.1 | NP_001257410.1 | 4, 5 | 491 | ||
| NM_002798.2 | NP_002789.1 | 4, 5 | 491 | |||
| GenBank mRNA | AK311768.1 | - | 4, 5 | 454 | ||
| BC000835.2 | - | 4, 5 | 458 | 3456009 | ||
| BF671774.1 | - | 4, 5 | 491 | |||
| BM553399.1 | - | 4, 5 | 438 | |||
| BQ691763.1 | - | 4, 5 | 473 | |||
| BT009809.1 | - | 4, 5 | 440 | |||
| BU848746.1 | - | 4, 5 | 423 | |||
| CR457065.1 | - | 4, 5 | 440 | |||

PSMB6
Genomic Map