siRNA Details
| Gene Symbol: UBE4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105562.2 | NP_001099032.1 | 28 | 5411 | ||
| NM_006048.4 | NP_006039.2 | 27 | 5024 | |||
| XM_005263422.1 | XP_005263479.1 | 29 | 5388 | |||
| XM_011540488.2 | XP_011538790.1 | 28 | 4687 | |||
| XM_017000035.1 | XP_016855524.1 | 26 | 4474 | |||
| XM_017000036.1 | XP_016855525.1 | 28 | 4688 | |||
| GenBank mRNA | AF043117.1 | - | 28 | 4808 | ||

UBE4B
Genomic Map