siRNA Details
| Gene Symbol: RAB32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006834.4 | NP_006825.1 | 3 | 773 | ||
| GenBank mRNA | AF498958.1 | - | 3 | 593 | ||
| BC015061.1 | - | 3 | 775 | 4280407 | ||
| BT020016.1 | - | 3 | 593 | |||
| U59878.1 | - | 3 | 548 | |||
| U71127.1 | - | 3 | 606 | |||

RAB32
Genomic Map