siRNA Details
| Gene Symbol: IFT27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177701.2 | NP_001171172.1 | 7 | 1023 | ||
| NM_006860.4 | NP_006851.1 | 7 | 1020 | |||
| XM_006724106.2 | XP_006724169.1 | 8 | 784 | |||
| XM_017028540.1 | XP_016884029.1 | 7 | 1027 | |||
| GenBank mRNA | AB209424.1 | - | 5 | 6414 | ||
| AK090708.1 | - | 1 | 3294 | |||
| AL022729.1 | - | 7 | 964 | |||
| BC000566.2 | - | 7 | 1001 | 3163341 | ||
| BX537634.1 | - | 1 | 3797 | |||

IFT27
Genomic Map