siRNA Details
| Gene Symbol: RAB35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167606.1 | NP_001161078.1 | 5 | 644 | ||
| NM_006861.6 | NP_006852.1 | 6 | 769 | |||
| XM_011537779.1 | XP_011536081.1 | 6 | 706 | |||
| XM_017018723.1 | XP_016874212.1 | 5 | 546 | |||
| GenBank mRNA | AF498960.1 | - | 6 | 580 | ||
| AK125543.1 | - | 7 | 1544 | |||
| AK298391.1 | - | 2 | 164 | |||
| AK304620.1 | - | 5 | 551 | |||
| AK312538.1 | - | 6 | 679 | |||
| BC015931.2 | - | 6 | 684 | 3907209 | ||
| BM467132.1 | - | 6 | 751 | |||
| BQ233709.1 | - | 6 | 725 | |||
| BT020024.1 | - | 6 | 580 | |||
| CR536486.1 | - | 6 | 580 | |||
| CR541683.1 | - | 6 | 580 | |||
| X79781.1 | - | 6 | 661 | |||

RAB35
Genomic Map