siRNA Details
| Gene Symbol: UBE2T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310326.1 | NP_001297255.1 | 7 | 703 | ||
| NM_014176.3 | NP_054895.1 | 7 | 699 | |||
| GenBank mRNA | AB032931.1 | - | 6 | 605 | ||
| AF160215.1 | - | 7 | 656 | |||
| AF161499.1 | - | 7 | 689 | |||
| AK000504.1 | - | 7 | 654 | |||
| AY542309.1 | - | 6 | 564 | |||
| BC004152.2 | - | 7 | 661 | 2821806 | ||
| BC019284.1 | - | 7 | 661 | 2821806 | ||
| BM460257.1 | - | 7 | 676 | |||

UBE2T
Genomic Map