siRNA Details
| Gene Symbol: RAB30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286059.1 | NP_001272988.1 | 4 | 613 | ||
| NM_001286060.1 | NP_001272989.1 | 4 | 448 | |||
| NM_001286061.1 | NP_001272990.1 | 4 | 346 | |||
| NM_014488.4 | NP_055303.2 | 5 | 549 | |||
| GenBank mRNA | AF498956.1 | - | 3 | 207 | ||
| AK291376.1 | - | 4 | 608 | |||
| AK310082.1 | - | 2 | 220 | |||
| AK312323.1 | - | 4 | 354 | |||
| BC014213.2 | - | 5 | 468 | 4763140 | ||
| BP336371.1 | - | 4 | 346 | |||
| BX641138.1 | - | 4 | 397 | |||
| CR542106.1 | - | 3 | 207 | |||
| DB151524.1 | - | 4 | 449 | |||
| U57092.1 | - | 3 | 225 | |||

RAB30
Genomic Map