siRNA Details
| Gene Symbol: USP34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014709.3 | NP_055524.3 | 6 | 819 | ||
| GenBank mRNA | AB011142.2 | - | 7 | 874 | ||
| AJ586138.1 | - | 4 | 344 | |||
| BC141839.1 | - | 7 | 874 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

USP34| Gene Symbol: USP34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014709.3 | NP_055524.3 | 6 | 819 | ||
| GenBank mRNA | AB011142.2 | - | 7 | 874 | ||
| AJ586138.1 | - | 4 | 344 | |||
| BC141839.1 | - | 7 | 874 | |||