siRNA Details
| Gene Symbol: RAB6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016577.3 | NP_057661.3 | 5 | 700 | ||
| XM_011512893.2 | XP_011511195.1 | 5 | 375 | |||
| GenBank mRNA | AF166492.1 | - | 5 | 656 | ||
| AF498940.1 | - | 5 | 319 | |||
| AK295451.1 | - | 6 | 437 | |||
| AK312378.1 | - | 5 | 620 | |||
| BC002510.2 | - | 5 | 669 | 3050592 | ||
| BC078662.1 | - | 5 | 448 | 6450986 | ||
| BT007263.1 | - | 5 | 319 | |||
| U66623.1 | - | 2 | 107 | |||

RAB6B
Genomic Map