siRNA Details
| Gene Symbol: RAB6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016577.3 | NP_057661.3 | 8 | 1101 | ||
| XM_011512893.2 | XP_011511195.1 | 8 | 776 | |||
| GenBank mRNA | AB073348.1 | - | 2 | 220 | ||
| AF166492.1 | - | 8 | 1057 | |||
| AK295451.1 | - | 9 | 838 | |||
| BC002510.2 | - | 8 | 1070 | 3050592 | ||
| BC078662.1 | - | 8 | 849 | 6450986 | ||

RAB6B
Genomic Map