siRNA Details
| Gene Symbol: RRAGB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006064.4 | NP_006055.3 | 8 | 1424 | ||
| NM_016656.3 | NP_057740.2 | 9 | 1508 | |||
| XM_005261973.4 | XP_005262030.1 | 9 | 845 | |||
| XM_017029217.1 | XP_016884706.1 | 8 | 754 | |||
| GenBank mRNA | AK289407.1 | - | 8 | 1357 | ||
| AL831926.1 | - | 8 | 1363 | |||
| BC034726.1 | - | 8 | 1355 | 3919252 | ||
| CD676755.1 | - | 2 | 149 | |||
| X90530.1 | - | 9 | 1299 | |||

RRAGB
Genomic Map