siRNA Details
| Gene Symbol: UBE2R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017811.3 | NP_060281.2 | 5 | 1690 | ||
| XM_005251496.2 | XP_005251553.1 | 4 | 1623 | |||
| XM_011517949.1 | XP_011516251.1 | 4 | 1575 | |||
| XM_017014860.1 | XP_016870349.1 | 3 | 1488 | |||
| GenBank mRNA | AJ240087.1 | - | 5 | 1120 | ||
| AK000426.1 | - | 5 | 1307 | |||
| AK225880.1 | - | 5 | 1295 | |||
| BC004862.2 | - | 5 | 1297 | 3838157 | ||
| BC047584.1 | - | 5 | 1544 | 4827644 | ||

UBE2R2
Genomic Map