siRNA Details
| Gene Symbol: RAB25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020387.3 | NP_065120.2 | 3 | 544 | ||
| NR_133653.1 | 2 | 348 | ||||
| GenBank mRNA | AF083124.1 | - | 3 | 505 | ||
| AF274025.1 | - | 3 | 523 | |||
| BC004416.1 | - | 1 | 34 | |||
| BC009831.2 | - | 3 | 518 | 3926839 | ||
| BC033322.1 | - | 3 | 508 | 4775725 | ||
| BP273992.1 | - | 2 | 333 | |||

RAB25
Genomic Map