siRNA Details
| Gene Symbol: PIK3C2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321378.1 | NP_001308307.1 | 28 | 4566 | ||
| NM_001321380.1 | NP_001308309.1 | 26 | 3367 | |||
| NM_002645.3 | NP_002636.2 | 27 | 4497 | |||
| XM_011520186.2 | XP_011518488.1 | 26 | 4324 | |||
| XM_017017924.1 | XP_016873413.1 | 28 | 4513 | |||
| GenBank mRNA | AK294416.1 | - | 26 | 3354 | ||
| AK309610.1 | - | 8 | 930 | |||
| BC113658.1 | - | 26 | 4349 | 8322710 | ||
| BX648778.1 | - | 17 | 2433 | |||
| Y13367.1 | - | 26 | 4283 | |||

PIK3C2A
Genomic Map