siRNA Details
| Gene Symbol: RAB18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256410.1 | NP_001243339.1 | 8 | 1492 | ||
| NM_001256411.1 | NP_001243340.1 | 6 | 1338 | |||
| NM_001256412.1 | NP_001243341.1 | 5 | 1213 | |||
| NM_001256415.1 | NP_001243344.1 | 6 | 1333 | |||
| NM_021252.4 | NP_067075.1 | 7 | 1405 | |||
| NR_046172.1 | 6 | 1343 | ||||
| GenBank mRNA | AB209698.1 | - | 3 | 6167 | ||
| AK001555.1 | - | 7 | 1209 | |||
| AK223153.1 | - | 7 | 1232 | |||
| AK295443.1 | - | 5 | 1083 | |||
| AK299457.1 | - | 6 | 1209 | |||
| AL136734.1 | - | 7 | 1258 | |||
| AY574034.1 | - | 8 | 1381 | |||
| BC015014.2 | - | 7 | 1259 | 3922029 | ||
| BC029350.1 | - | 7 | 1275 | 4819866 | ||

RAB18
Genomic Map