siRNA Details
| Gene Symbol: PIK3CG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282426.1 | NP_001269355.1 | 11 | 4728 | ||
| NM_001282427.1 | NP_001269356.1 | 11 | 4652 | |||
| NM_002649.3 | NP_002640.2 | 11 | 4877 | |||
| XM_005250443.3 | XP_005250500.1 | 11 | 4745 | |||
| GenBank mRNA | BX648341.1 | - | 11 | 4710 | ||
| X83368.1 | - | 11 | 4888 | |||

PIK3CG
Genomic Map